| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100030560.mm.1 |
NM_001042565.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030560.mm.1 |
NM_001042565.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030560.mm.1 |
TC1100003318.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100030558.mm.1 |
NM_001042565.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030558.mm.1 |
NM_001042565.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030558.mm.1 |
TC1100003318.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100030556.mm.1 |
NM_001042565.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030556.mm.1 |
NM_001042565.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030556.mm.1 |
NM_019653.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030556.mm.1 |
NM_019653.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030556.mm.1 |
TC1100003318.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100030559.mm.1 |
NM_001042565.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030559.mm.1 |
NM_001042565.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100030559.mm.1 |
TC1100003318.mm.1 |
MTA TC |