Assay Details
| Gene Symbol: Tubd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199045.1 | NP_001185974.1 | 3 - 4 | 708 | 135 | |
| NM_001199046.1 | NP_001185975.1 | 3 - 4 | 602 | 135 | ||
| NM_001199047.1 | NP_001185976.1 | 3 - 4 | 708 | 135 | ||
| NM_019756.3 | NP_062730.1 | 3 - 4 | 602 | 135 | ||
| GenBank mRNA | AB024329.1 | - | 2 - 3 | 376 | 135 | |
| AB031043.1 | - | 2 - 3 | 314 | 135 | ||
| AF081568.1 | - | 3 - 4 | 442 | 135 | ||
| AK016086.1 | - | 3 - 4 | 402 | 135 | ||
| AK030214.1 | - | 3 - 4 | 708 | 135 | ||
| AK169454.1 | - | 3 - 4 | 603 | 135 | ||
| AK170304.1 | - | 3 - 4 | 567 | 135 | ||
| AK171546.1 | - | 3 - 4 | 569 | 135 | ||
| BC026465.1 | - | 3 - 4 | 585 | 4484099 | 135 | |
| BC132325.1 | - | 3 - 4 | 435 | 40130602 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubd1
Genomic Map