Assay Details
| Gene Symbol: Tubd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199045.1 | NP_001185974.1 | 6 - 7 | 1256 | 67 | |
| NM_001199046.1 | NP_001185975.1 | 6 - 7 | 1150 | 67 | ||
| GenBank mRNA | AB031043.1 | - | 5 - 6 | 862 | 67 | |
| AK030214.1 | - | 6 - 7 | 1256 | 67 | ||
More Information
Set Membership: |
|

Tubd1
Genomic Map