Assay Details
| Gene Symbol: Pctp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008796.2 | NP_032822.2 | 4 - 5 | 561 | 63 | |
| GenBank mRNA | AF114437.1 | - | 2 - 3 | 266 | 63 | |
| AF151639.1 | - | 4 - 5 | 532 | 63 | ||
| BB614866.1 | - | 409 | 63 | |||
| Z50024.1 | - | 4 - 5 | 390 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pctp
Genomic Map