Assay Details
| Gene Symbol: Mrps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291270.1 | NP_001278199.1 | 4 - 5 | 461 | 91 | |
| NM_001291271.1 | NP_001278200.1 | 4 - 5 | 705 | 91 | ||
| NM_001291272.1 | NP_001278201.1 | 4 - 5 | 572 | 91 | ||
| NM_001291273.1 | NP_001278202.1 | 3 - 4 | 306 | 91 | ||
| NM_024174.6 | NP_077136.2 | 4 - 5 | 477 | 91 | ||
| NR_111902.1 | 3 - 4 | 401 | 91 | |||
| GenBank mRNA | AK009880.1 | - | 4 - 5 | 532 | 91 | |
| AK014375.2 | - | 4 - 5 | 447 | 91 | ||
| AK168051.1 | - | 4 - 5 | 671 | 91 | ||
| AK168277.1 | - | 4 - 5 | 432 | 91 | ||
| BC002155.1 | - | 4 - 5 | 524 | 91 | ||
| BC019980.1 | - | 4 - 5 | 447 | 4458402 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998475 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010858 | BC019980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010858 | BC002155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010858 | AK168051.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010858 | AK168277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010861 | NM_024174.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493299 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150410 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010858 | BC019980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010858 | BC002155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010858 | AK168051.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010858 | AK168277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010861 | NM_024174.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338542 | BC019980.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338542 | BC002155.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338542 | AK009880.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338542 | AK014375.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338542 | NM_024174.5 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27998450 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150410 | - | - |
| Whole Mouse Genome G4122A | A_51_P338542 | BC019980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338542 | BC002155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338542 | AK009880.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338542 | AK014375.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338542 | NM_024174.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5420 | BC019980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5420 | BC002155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5420 | AK009880.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5420 | AK014375.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5420 | NM_024174.5 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103918 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09689548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09689546 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09039461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09689549 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09039463 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27998475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27998550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09435178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09538802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09101424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09106422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09101436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09358372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09252179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09252180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09376398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09345443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09538805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09409948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09149924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09727321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27998461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09003031 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380241 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421875_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421874_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459806_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380238 | - | - |
| Mouse Genome 430 2.0 Array | 1459806_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1421874_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421875_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421874_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1459806_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1421875_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64656_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64656_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100012888.mm.1 | NM_024174.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012888.mm.1 | TC1100001369.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012889.mm.1 | NM_024174.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012889.mm.1 | TC1100001369.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421874_a_at | - | - |
| Mouse Expression Set 430 | 1421875_a_at | - | - |
| Mouse Expression Set 430 | 1421874_PM_a_at | - | - |
| Mouse Expression Set 430 | 1421875_PM_a_at | - | - |
| Mouse Expression Set 430 | 1459806_x_at | - | - |
| Mouse Expression Set 430 | 1459806_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421874_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421875_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421874_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421875_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421874_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421874_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421875_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421875_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459806_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459806_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 545219 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3610551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2762361 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2762361 | - | - |
| mouse-6 v1.1 (Illumina) | scl064656.5_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl064656.5_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2909068 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2909068 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2762361 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl064656.5_1-S | - | - |

Mrps23
Genomic Map