Assay Details
| Gene Symbol: Mapk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168508.1 | NP_001161980.1 | 9 - 10 | 1166 | 148 | |
| GenBank mRNA | AF195850.1 | - | 6 - 7 | 489 | 148 | |
| AK089059.1 | - | 9 - 10 | 1080 | 148 | ||
| BC085303.1 | - | 9 - 10 | 1142 | 5389457 | 148 | |
| CX568915.1 | - | 8 - 9 | 560 | 148 | ||
| D83073.1 | - | 9 - 10 | 997 | 148 | ||
More Information
Set Membership: |
|

Mapk14
Genomic Map