Assay Details
| Gene Symbol: Atp5g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161419.1 | NP_001154891.1 | 4 - 5 | 376 | 67 | |
| NM_007506.6 | NP_031532.2 | 4 - 5 | 369 | 67 | ||
| GenBank mRNA | AK008191.1 | - | 4 - 5 | 365 | 67 | |
| AK008998.1 | - | 4 - 5 | 320 | 67 | ||
| AK009883.2 | - | 4 - 5 | 365 | 67 | ||
| AK010868.1 | - | 4 - 5 | 338 | 67 | ||
| AK011054.1 | - | 4 - 5 | 330 | 67 | ||
| AK011432.1 | - | 4 - 5 | 319 | 67 | ||
| AK167884.1 | - | 4 - 5 | 338 | 67 | ||
| BC003854.1 | - | 4 - 5 | 349 | 3601516 | 67 | |
| BC094664.1 | - | 306 | 30532297 | 67 | ||
| CF749235.1 | - | 1 - 2 | 114 | 67 | ||
| L19737.1 | - | 3 - 4 | 296 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28042294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28042297 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28042295 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1956064 | BC094664.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816117 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178880 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001683 | AK011054.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001683 | AK167884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001683 | AK009883.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001683 | AK010868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001683 | BC003854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1956064 | BC094664.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183292 | AK011054.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183292 | AK011432.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183292 | AK167884.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183292 | AK008998.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183292 | AK009883.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28042289 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02402013 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178880 | - | - |
| Whole Mouse Genome G4122A | A_51_P183292 | AK011054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183292 | AK011432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183292 | AK167884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183292 | AK008998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183292 | AK009883.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102484 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135204 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178880 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466672 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701192 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701183 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701184 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701190 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28042295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28042297 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28042294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28042279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09701192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09177529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09115759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09702922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09177526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09069771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28042274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09177523 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10391102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390241 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390238 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10391101 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444874_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416020_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10391100 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390237 | - | - |
| Mouse Genome 430 2.0 Array | 1444874_at | - | - |
| Mouse Genome 430 2.0 Array | 1444874_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416020_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416020_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11951_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11951_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100033075.mm.1 | NM_001161419.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033075.mm.1 | NM_001161419.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033075.mm.1 | NM_007506.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033075.mm.1 | NM_007506.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033075.mm.1 | TC1100003610.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100033072.mm.1 | NM_001161419.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033072.mm.1 | NM_001161419.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033072.mm.1 | NM_007506.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033072.mm.1 | NM_007506.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033072.mm.1 | TC1100003610.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100033071.mm.1 | TC1100003610.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416020_a_at | - | - |
| Mouse Expression Set 430 | 1416020_PM_a_at | - | - |
| Mouse Expression Set 430 | 1444874_PM_at | - | - |
| Mouse Expression Set 430 | 1444874_at | - | - |
| Murine Genome U74v2 Set | 161324_r_at | - | - |
| Murine Genome U74v2 Set | 96032_PM_at | - | - |
| Murine Genome U74v2 Set | 161324_PM_r_at | - | - |
| Murine Genome U74v2 Set | 96032_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416020_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416020_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444874_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444874_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5290019 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5290019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005290019 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230991 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230991 | - | - |
| mouse-6 v1.1 (Illumina) | scl011951.2_99-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5290019 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011951.2_99-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230991 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230991 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl011951.2_99-S | - | - |

Atp5g1
Genomic Map