Assay Details
| Gene Symbol: Glycam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289587.1 | NP_001276516.1 | 1 - 2 | 115 | 66 | |
| NM_008134.3 | NP_032160.1 | 1 - 2 | 115 | 66 | ||
| GenBank mRNA | AK021358.1 | - | 1 - 2 | 113 | 66 | |
| BC145548.1 | - | 1 - 2 | 88 | 9054112 | 66 | |
| BC145832.1 | - | 1 - 2 | 88 | 40131135 | 66 | |
| BC145858.1 | - | 1 - 2 | 88 | 40131161 | 66 | |
| M93428.1 | - | 1 - 2 | 87 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glycam1
Genomic Map