Assay Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF401483.1 | - | 1 - 2 | 50 | 96 | |
| AK045256.1 | - | 1 - 2 | 37 | 96 | ||
| AK158718.1 | - | 1 - 2 | 110 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF401483.1, AK045256.1 transcripts.
Set Membership: |
|

Letmd1
Genomic Map