Assay Details
| Gene Symbol: Med1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080118.1 | NP_001073587.1 | 7 - 8 | 712 | 135 | |
| NM_013634.2 | NP_038662.2 | 7 - 8 | 577 | 135 | ||
| NM_134027.2 | NP_598788.2 | 7 - 8 | 712 | 135 | ||
| GenBank mRNA | AF000294.1 | - | 7 - 8 | 577 | 135 | |
| AF332073.1 | - | 6 - 7 | 457 | 135 | ||
| AF332074.1 | - | 6 - 7 | 457 | 135 | ||
| AK005207.1 | - | 7 - 8 | 694 | 134 | ||
| AK049203.1 | - | 7 - 8 | 703 | 135 | ||
| AK054437.1 | - | 7 - 8 | 713 | 135 | ||
| AK147199.1 | - | 7 - 8 | 689 | 135 | ||
| BC079636.1 | - | 7 - 8 | 682 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med1
Genomic Map