Assay Details
| Gene Symbol: Med1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080118.1 | NP_001073587.1 | 9 - 10 | 856 | 102 | |
| NM_013634.2 | NP_038662.2 | 9 - 10 | 721 | 102 | ||
| NM_134027.2 | NP_598788.2 | 9 - 10 | 856 | 102 | ||
| GenBank mRNA | AF332073.1 | - | 8 - 9 | 601 | 102 | |
| AF332074.1 | - | 8 - 9 | 601 | 102 | ||
| AK049203.1 | - | 9 - 10 | 847 | 102 | ||
| AK054437.1 | - | 9 - 10 | 857 | 102 | ||
| AK147199.1 | - | 9 - 10 | 833 | 102 | ||
| BC079636.1 | - | 9 - 10 | 826 | 102 | ||
More Information
Set Membership: |
|

Med1
Genomic Map