Assay Details
| Gene Symbol: Sbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081030.2 | NP_001074499.2 | 32 - 33 | 4675 | 93 | |
| NM_001170561.1 | NP_001164032.1 | 33 - 34 | 4753 | 93 | ||
| GenBank mRNA | AK020972.1 | - | 3 - 4 | 316 | 93 | |
| AK129485.1 | - | 32 - 33 | 4619 | 93 | ||
| AK147332.1 | - | 32 - 33 | 4632 | 93 | ||
| BC029156.1 | - | 3 - 4 | 311 | 93 | ||
| BC098209.1 | - | 13 - 14 | 2018 | 93 | ||
| BC157935.1 | - | 32 - 33 | 4565 | 9088115 | 93 | |
| BC158013.1 | - | 33 - 34 | 4643 | 9007320 | 93 | |
| BC172094.1 | - | 32 - 33 | 4565 | 9054380 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC029156.1 transcripts.
Set Membership: |
|

Sbf1
Genomic Map