Assay Details
| Gene Symbol: Sbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081030.2 | NP_001074499.2 | 37 - 38 | 5481 | 84 | |
| NM_001170561.1 | NP_001164032.1 | 38 - 39 | 5559 | 84 | ||
| GenBank mRNA | AK076039.1 | - | 5 - 6 | 726 | 84 | |
| AK129485.1 | - | 37 - 38 | 5425 | 84 | ||
| AK147332.1 | - | 37 - 38 | 5438 | 84 | ||
| BC029156.1 | - | 8 - 9 | 1117 | 84 | ||
| BC098209.1 | - | 18 - 19 | 2824 | 84 | ||
| BC157935.1 | - | 37 - 38 | 5371 | 9088115 | 84 | |
| BC158013.1 | - | 38 - 39 | 5449 | 9007320 | 84 | |
| BC172094.1 | - | 37 - 38 | 5371 | 9054380 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sbf1
Genomic Map