Assay Details
| Gene Symbol: Ptger4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136079.2 | NP_001129551.1 | 1 - 2 | 571 | 60 | |
| GenBank mRNA | BC009023.1 | - | 1 - 2 | 66 | 3481696 | 60 |
| BC011193.1 | - | 1 - 2 | 556 | 4037704 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC009023.1, BC011193.1, NM_001136079.2 transcripts.
Set Membership: |
|

Ptger4
Genomic Map