Assay Details
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048178.2 | NP_001041643.1 | 7 - 8 | 1005 | 157 | |
NM_001286370.1 | NP_001273299.1 | 7 - 8 | 706 | 157 | ||
NM_010284.3 | NP_034414.2 | 7 - 8 | 1005 | 157 | ||
GenBank mRNA | AI194700.1 | - | 2 - 3 | 157 | ||
AK143886.1 | - | 7 - 8 | 707 | 157 | ||
AK143925.1 | - | 7 - 8 | 803 | 157 | ||
BC024375.1 | - | 7 - 8 | 840 | 157 | ||
BC056645.1 | - | 7 - 8 | 821 | 157 | ||
BC075720.1 | - | 7 - 8 | 853 | 5044060 | 157 | |
M31680.1 | - | 7 - 8 | 741 | 157 | ||
M33324.1 | - | 7 - 8 | 741 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
