Assay Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039181.1 | NP_001034270.1 | 3 - 4 | 1078 | 84 | |
| NM_001286395.1 | NP_001273324.1 | 4 - 5 | 694 | 84 | ||
| NM_008728.2 | NP_032754.2 | 3 - 4 | 1078 | 84 | ||
| GenBank mRNA | AK079196.1 | - | 4 - 5 | 611 | 84 | |
| AK166535.1 | - | 2 - 3 | 307 | 84 | ||
| BC055897.1 | - | 3 - 4 | 1078 | 4019152 | 84 | |
| BQ832991.1 | - | 2 - 3 | 247 | 84 | ||
| D78175.1 | - | 3 - 4 | 1043 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK166535.1 transcripts.
Set Membership: |
|

Npr3
Genomic Map