Assay Details
| Gene Symbol: Skp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285980.1 | NP_001272909.1 | 2 - 3 | 473 | 73 | |
| NM_013787.3 | NP_038815.1 | 2 - 3 | 543 | 73 | ||
| GenBank mRNA | AF083215.1 | - | 2 - 3 | 406 | 73 | |
| AK006221.1 | - | 2 - 3 | 473 | 73 | ||
| AK032979.1 | - | 2 - 3 | 454 | 73 | ||
| AK044212.1 | - | 2 - 3 | 444 | 73 | ||
| AK166034.1 | - | 2 - 3 | 444 | 73 | ||
| AK167677.1 | - | 2 - 3 | 441 | 73 | ||
| BC003468.1 | - | 2 - 3 | 416 | 73 | ||
| BC092236.1 | - | 2 - 3 | 417 | 30610417 | 73 | |
| BC098371.1 | - | 2 - 3 | 526 | 73 | ||
| BY740905.1 | - | 2 - 3 | 544 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Skp2
Genomic Map