Assay Details
| Gene Symbol: Arhgap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164627.1 | NP_001158099.1 | 7 - 8 | 602 | 78 | |
| NM_001164628.1 | NP_001158100.1 | 6 - 7 | 594 | 78 | ||
| NM_001205334.1 | NP_001192263.1 | 7 - 8 | 622 | 78 | ||
| NM_028455.4 | NP_082731.2 | 6 - 7 | 614 | 78 | ||
| GenBank mRNA | AK040142.1 | - | 6 - 7 | 564 | 78 | |
| AK086816.1 | - | 6 - 7 | 568 | 78 | ||
| AK088242.1 | - | 6 - 7 | 570 | 78 | ||
| BC005563.1 | - | 5 - 6 | 557 | 3965919 | 78 | |
| BC010306.1 | - | 6 - 7 | 584 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap8
Genomic Map