Assay Details
| Gene Symbol: Cpne8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025815.2 | NP_080091.1 | 11 - 12 | 927 | 121 | |
| GenBank mRNA | AK004559.1 | - | 11 - 12 | 915 | 121 | |
| BC017626.1 | - | 10 - 11 | 821 | 4502073 | 121 | |
| BC138745.1 | - | 11 - 12 | 926 | 8861767 | 121 | |
| BC138746.1 | - | 11 - 12 | 926 | 8861768 | 121 | |
More Information
Set Membership: |
|

Cpne8
Genomic Map