Assay Details
| Gene Symbol: Cpne8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025815.2 | NP_080091.1 | 13 - 14 | 1037 | 72 | |
| GenBank mRNA | AK004559.1 | - | 13 - 14 | 1025 | 72 | |
| BC017626.1 | - | 12 - 13 | 931 | 4502073 | 72 | |
| BC138745.1 | - | 13 - 14 | 1036 | 8861767 | 72 | |
| BC138746.1 | - | 13 - 14 | 1036 | 8861768 | 72 | |
More Information
Set Membership: |
|

Cpne8
Genomic Map