Assay Details
| Gene Symbol: Cpne8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025815.2 | NP_080091.1 | 4 - 5 | 403 | 146 | |
| GenBank mRNA | AK004559.1 | - | 4 - 5 | 391 | 146 | |
| AK010227.1 | - | 4 - 5 | 403 | 146 | ||
| AK013407.1 | - | 4 - 5 | 388 | 146 | ||
| BC017626.1 | - | 4 - 5 | 374 | 4502073 | 146 | |
| BC138745.1 | - | 4 - 5 | 402 | 8861767 | 146 | |
| BC138746.1 | - | 4 - 5 | 402 | 8861768 | 146 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC017626.1 transcripts.
Set Membership: |
|

Cpne8
Genomic Map