Assay Details
| Gene Symbol: Poldip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178627.3 | NP_848742.1 | 2 - 3 | 535 | 77 | |
| GenBank mRNA | AK052722.1 | - | 2 - 3 | 530 | 77 | |
| AK081139.1 | - | 2 - 3 | 495 | 77 | ||
| AK083924.1 | - | 2 - 3 | 536 | 77 | ||
| AK087446.1 | - | 2 - 3 | 544 | 77 | ||
| AK152060.1 | - | 2 - 3 | 576 | 77 | ||
| AK167552.1 | - | 2 - 3 | 479 | 77 | ||
| AK194128.1 | - | 1 - 2 | 52 | 77 | ||
| AK201338.1 | - | 1 - 2 | 52 | 77 | ||
| AK220221.1 | - | 2 - 3 | 551 | 77 | ||
| BC037652.1 | - | 2 - 3 | 520 | 5136112 | 77 | |
| BC046505.1 | - | 2 - 3 | 464 | 4912042 | 77 | |
| BC058225.1 | - | 2 - 3 | 531 | 6307930 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poldip3
Genomic Map