Assay Details
| Gene Symbol: Ttll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178869.4 | NP_849200.2 | 9 - 10 | 1279 | 61 | |
| GenBank mRNA | AK031917.1 | - | 9 - 10 | 1250 | 61 | |
| AK047046.1 | - | 9 - 10 | 1264 | 61 | ||
| AK140472.1 | - | 9 - 10 | 1247 | 61 | ||
| AK168786.1 | - | 9 - 10 | 1250 | 61 | ||
| AK169150.1 | - | 9 - 10 | 1266 | 61 | ||
| BC010510.1 | - | 9 - 10 | 1228 | 4191464 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll1
Genomic Map