Assay Details
| Gene Symbol: Ttll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178869.4 | NP_849200.2 | 5 - 6 | 706 | 107 | |
| GenBank mRNA | AK031917.1 | - | 5 - 6 | 677 | 107 | |
| AK047046.1 | - | 5 - 6 | 691 | 107 | ||
| AK084005.1 | - | 5 - 6 | 674 | 107 | ||
| AK140472.1 | - | 5 - 6 | 674 | 107 | ||
| AK168786.1 | - | 5 - 6 | 677 | 107 | ||
| AK169150.1 | - | 5 - 6 | 693 | 107 | ||
| BC010510.1 | - | 5 - 6 | 655 | 4191464 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll1
Genomic Map