Assay Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175543.3 | NP_780752.1 | 10 - 11 | 1336 | 76 | |
| GenBank mRNA | AK046734.1 | - | 10 - 11 | 1338 | 76 | |
| AK122559.1 | - | 2 - 3 | 172 | 76 | ||
| AK147219.1 | - | 10 - 11 | 1407 | 76 | ||
| AK147344.1 | - | 10 - 11 | 1532 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip4
Genomic Map