Assay Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175543.3 | NP_780752.1 | 9 - 10 | 1201 | 73 | |
| GenBank mRNA | AK046734.1 | - | 9 - 10 | 1203 | 73 | |
| AK147219.1 | - | 9 - 10 | 1272 | 73 | ||
| AK147344.1 | - | 9 - 10 | 1397 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip4
Genomic Map