Assay Details
| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277106.1 | NP_001264035.1 | 4 - 5 | 679 | 68 | |
| NM_018776.2 | NP_061246.1 | 4 - 5 | 679 | 68 | ||
| GenBank mRNA | AF046060.1 | - | 4 - 5 | 654 | 68 | |
| AF120152.1 | - | 4 - 5 | 588 | 68 | ||
| AK037880.1 | - | 4 - 5 | 627 | 68 | ||
| AK078569.1 | - | 4 - 5 | 624 | 68 | ||
| AK137528.1 | - | 4 - 5 | 631 | 68 | ||
| BC004780.1 | - | 4 - 5 | 626 | 3257685 | 68 | |
More Information
Set Membership: |
|

Crlf3
Genomic Map