Assay Details
| Gene Symbol: Trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009546.2 | NP_033572.2 | 3 - 4 | 1008 | 82 | |
| GenBank mRNA | AK154533.1 | - | 3 - 4 | 1001 | 82 | |
| AK160898.1 | - | 3 - 4 | 960 | 82 | ||
| AK169562.1 | - | 3 - 4 | 1009 | 82 | ||
| BC006908.1 | - | 3 - 4 | 977 | 2651664 | 82 | |
| BC034276.1 | - | 3 - 4 | 980 | 4022161 | 82 | |
| D63902.1 | - | 3 - 4 | 931 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim25
Genomic Map