Assay Details
| Gene Symbol: Sept6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253706.1 | NP_001240635.1 | 10 - 11 | 1614 | 68 | |
| GenBank mRNA | AK172894.1 | - | 10 - 11 | 1395 | 68 | |
| BC010489.1 | - | 10 - 11 | 1400 | 4168214 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept6
Genomic Map