Assay Details
| Gene Symbol: Sept6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253706.1 | NP_001240635.1 | 9 - 10 | 1573 | 73 | |
| GenBank mRNA | AK172894.1 | - | 9 - 10 | 1354 | 73 | |
| BC010489.1 | - | 9 - 10 | 1359 | 4168214 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept6
Genomic Map