Assay Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243132.1 | NP_001230061.1 | 1 - 2 | 103 | 82 | |
| GenBank mRNA | AK020982.1 | - | 1 - 2 | 103 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan2
Genomic Map