Assay Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243132.1 | NP_001230061.1 | 4 - 5 | 370 | 97 | |
| NM_027533.3 | NP_081809.2 | 4 - 5 | 565 | 97 | ||
| GenBank mRNA | AK018178.1 | - | 4 - 5 | 368 | 97 | |
| AK020982.1 | - | 4 - 5 | 370 | 97 | ||
| AK031683.1 | - | 3 - 4 | 274 | 97 | ||
| AK032520.1 | - | 4 - 5 | 391 | 97 | ||
| AK049888.1 | - | 4 - 5 | 367 | 97 | ||
| AK079560.1 | - | 4 - 5 | 369 | 97 | ||
| BB642061.1 | - | 4 - 5 | 565 | 96 | ||
| BC007185.1 | - | 4 - 5 | 806 | 3591472 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan2
Genomic Map