Assay Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243132.1 | NP_001230061.1 | 6 - 7 | 546 | 108 | |
| NM_027533.3 | NP_081809.2 | 6 - 7 | 741 | 108 | ||
| GenBank mRNA | AK018178.1 | - | 6 - 7 | 544 | 108 | |
| AK020982.1 | - | 6 - 7 | 546 | 108 | ||
| AK031683.1 | - | 5 - 6 | 450 | 108 | ||
| AK032520.1 | - | 6 - 7 | 567 | 108 | ||
| AK049888.1 | - | 6 - 7 | 543 | 108 | ||
| AK079560.1 | - | 6 - 7 | 545 | 108 | ||
| BC007185.1 | - | 6 - 7 | 982 | 3591472 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan2
Genomic Map