Assay Details
| Gene Symbol: Tsc22d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010286.4 | NP_034416.3 | 1 - 2 | 342 | 89 | |
| GenBank mRNA | AF024519.1 | - | 1 - 2 | 327 | 89 | |
| AF201288.1 | - | 1 - 2 | 322 | 89 | ||
| AK083389.1 | - | 1 - 2 | 341 | 89 | ||
| AK144196.1 | - | 1 - 2 | 345 | 89 | ||
| BC028813.1 | - | 1 - 2 | 332 | 5136440 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tsc22d3
Genomic Map