Assay Details
| Gene Symbol: Ptgr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252626.1 | NP_001239555.1 | 4 - 5 | 516 | 170 | |
| GenBank mRNA | AK007522.1 | - | 4 - 5 | 399 | 170 | |
| AK040396.1 | - | 4 - 5 | 448 | 170 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252626.1 transcripts.
Set Membership: |
|

Ptgr2
Genomic Map