Assay Details
| Gene Symbol: Ptgr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252625.1 | NP_001239554.1 | 4 - 5 | 639 | 103 | |
| NM_001252626.1 | NP_001239555.1 | 5 - 6 | 711 | 103 | ||
| NM_029880.3 | NP_084156.2 | 4 - 5 | 582 | 103 | ||
| GenBank mRNA | AK007522.1 | - | 5 - 6 | 595 | 103 | |
| AK021033.1 | - | 4 - 5 | 474 | 103 | ||
| AK036168.1 | - | 4 - 5 | 504 | 103 | ||
| AK040396.1 | - | 5 - 6 | 643 | 103 | ||
| AK159187.1 | - | 4 - 5 | 478 | 103 | ||
| AK159932.1 | - | 4 - 5 | 479 | 103 | ||
| AK168895.1 | - | 4 - 5 | 473 | 103 | ||
More Information
Set Membership: |
|

Ptgr2
Genomic Map