Assay Details
| Gene Symbol: Ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144828.1 | NP_659077.1 | 4 - 5 | 601 | 78 | |
| GenBank mRNA | AB221544.1 | - | 4 - 5 | 250 | 78 | |
| AK141563.1 | - | 2 - 3 | 126 | 78 | ||
| AY640620.1 | - | 4 - 5 | 279 | 78 | ||
| AY640621.1 | - | 3 - 4 | 737 | 78 | ||
| BC011122.1 | - | 4 - 5 | 511 | 4164563 | 78 | |
| BC026568.1 | - | 4 - 5 | 601 | 4982294 | 78 | |
| BC031129.1 | - | 4 - 5 | 349 | 4988284 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r1b
Genomic Map