Assay Details
| Gene Symbol: Ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144828.1 | NP_659077.1 | 1 - 2 | 436 | 119 | |
| GenBank mRNA | AB221544.1 | - | 1 - 2 | 85 | 119 | |
| BC011122.1 | - | 1 - 2 | 346 | 4164563 | 119 | |
| BC026568.1 | - | 1 - 2 | 436 | 4982294 | 119 | |
| BC031129.1 | - | 1 - 2 | 184 | 4988284 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r1b
Genomic Map