Assay Details
| Gene Symbol: Gngt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284393.1 | NP_001271322.1 | 1 - 2 | 130 | 64 | |
| NM_023121.2 | NP_075610.1 | 1 - 2 | 130 | 64 | ||
| GenBank mRNA | BE847751.1 | - | 1 - 2 | 116 | 64 | |
| BY117112.1 | - | 1 - 2 | 130 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001284393.1, NM_023121.2 transcripts.
Set Membership: |
|

Gngt2
Genomic Map