Assay Details
| Gene Symbol: Atp6v1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023721.2 | NP_076210.1 | 8 - 9 | 779 | 120 | |
| GenBank mRNA | AF298810.1 | - | 8 - 9 | 600 | 120 | |
| AK146129.1 | - | 6 - 7 | 450 | 120 | ||
| AK146448.1 | - | 8 - 9 | 682 | 120 | ||
| AK147910.1 | - | 696 | 120 | |||
| AK152367.1 | - | 8 - 9 | 668 | 120 | ||
| AK165761.1 | - | 8 - 9 | 673 | 120 | ||
| BC033457.1 | - | 8 - 9 | 641 | 3991107 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566337 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566342 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566338 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566341 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904668 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10395 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91579 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180423 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165932 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28566319 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165932 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180423 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07676763 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07676742 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07676793 | - | - |
| Whole Mouse Genome G4122A | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK150981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK152367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK153250.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK165761.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393598 | AK146448.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107124 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165932 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180423 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07676742 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09870316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09870318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09870310 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09870305 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09870326 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09870333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566337 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566338 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28566342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28566271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10192097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09870333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10048418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10112944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10257025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28566313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10401141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401139 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401142 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401140 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401148 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416951_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438993_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416952_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10401138 | - | - |
| Mouse Genome 430 2.0 Array | 1416951_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416951_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1438993_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416952_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438993_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416952_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73834_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73834_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200013503.mm.1 | NM_023721.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013503.mm.1 | NM_023721.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013503.mm.1 | TC1200001971.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416951_a_at | - | - |
| Mouse Expression Set 430 | 1416952_PM_at | - | - |
| Mouse Expression Set 430 | 1438993_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416952_at | - | - |
| Mouse Expression Set 430 | 1416951_PM_a_at | - | - |
| Mouse Expression Set 430 | 1438993_a_at | - | - |
| Murine Genome U74v2 Set | 96951_PM_at | - | - |
| Murine Genome U74v2 Set | 96951_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416952_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416951_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416952_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438993_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438993_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416951_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416951_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438993_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438993_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416951_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416952_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416952_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2120037 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2120037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002120037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2647048 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2647048 | - | - |
| mouse-6 v1.1 (Illumina) | scl0073834.1_147-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2120037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0073834.1_147-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2647048 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2647048 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0073834.1_147-S | - | - |

Atp6v1d
Genomic Map