Assay Details
| Gene Symbol: Eif2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026114.3 | NP_080390.1 | 5 - 6 | 688 | 68 | |
| GenBank mRNA | AK009934.1 | - | 1 - 2 | 89 | 68 | |
| AK010592.1 | - | 5 - 6 | 718 | 68 | ||
| AK143499.1 | - | 5 - 6 | 689 | 68 | ||
| AK167763.1 | - | 5 - 6 | 690 | 68 | ||
| BC005463.1 | - | 5 - 6 | 668 | 3591411 | 68 | |
| BC016448.1 | - | 5 - 6 | 692 | 4234809 | 68 | |
| BC016497.1 | - | 5 - 6 | 692 | 4501321 | 68 | |
More Information
Set Membership: |
|

Eif2s1
Genomic Map