Assay Details
| Gene Symbol: Smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK151325.1 | - | 1 - 2 | 192 | 94 | |
| AK152795.1 | - | 1 - 2 | 192 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smdt1| Gene Symbol: Smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK151325.1 | - | 1 - 2 | 192 | 94 | |
| AK152795.1 | - | 1 - 2 | 192 | 94 | ||
Set Membership: |
|