Assay Details
| Gene Symbol: Krt24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029393.1 | NP_083669.1 | 5 - 6 | 1168 | 60 | |
| GenBank mRNA | AK009986.1 | - | 4 - 5 | 513 | 60 | |
| AK010165.1 | - | 5 - 6 | 1191 | 60 | ||
| AK053678.1 | - | 5 - 6 | 914 | 60 | ||
| BC129847.1 | - | 5 - 6 | 1168 | 40107361 | 60 | |
| BC129848.1 | - | 5 - 6 | 1168 | 40107363 | 60 | |
| GU046702.1 | - | 4 - 5 | 1232 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt24
Genomic Map