Assay Details
| Gene Symbol: Mill2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153761.3 | NP_715642.2 | 2 - 3 | 173 | 88 | |
| GenBank mRNA | AB086267.1 | - | 2 - 3 | 173 | 88 | |
| BC144886.1 | - | 2 - 3 | 171 | 9053434 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mill2
Genomic Map