| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100011164.mm.1 |
NR_038011.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011164.mm.1 |
NR_038011.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011164.mm.1 |
TC0100001348.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011162.mm.1 |
NR_038011.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011162.mm.1 |
NR_038011.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011162.mm.1 |
TC0100001348.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011160.mm.1 |
NR_038011.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011160.mm.1 |
NR_038011.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011160.mm.1 |
TC0100001348.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011159.mm.1 |
NR_038011.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011159.mm.1 |
NR_038011.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011159.mm.1 |
TC0100001348.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011168.mm.1 |
NR_038011.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011168.mm.1 |
NR_038011.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011168.mm.1 |
TC0100001348.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011170.mm.1 |
NR_038011.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011170.mm.1 |
NR_038011.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011170.mm.1 |
TC0100001348.mm.1 |
MTA TC |