Assay Details
| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102565.1 | NP_001096035.1 | 4 - 5 | 558 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alkbh1| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102565.1 | NP_001096035.1 | 4 - 5 | 558 | 78 | |
Set Membership: |
|