Assay Details
| Gene Symbol: Nhlh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178777.3 | NP_848892.1 | 2 - 3 | 508 | 102 | |
| GenBank mRNA | AK031747.1 | - | 2 - 3 | 510 | 102 | |
| AK053046.1 | - | 2 - 3 | 499 | 102 | ||
| AK053837.1 | - | 1 - 2 | 917 | 102 | ||
| AK079121.1 | - | 1 - 2 | 359 | 102 | ||
| AK084160.1 | - | 2 - 3 | 498 | 102 | ||
| AK084308.1 | - | 2 - 3 | 499 | 102 | ||
| AK140559.1 | - | 1 - 2 | 595 | 102 | ||
| BC058413.1 | - | 2 - 3 | 376 | 6816900 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031747.1, AK053837.1, AK079121.1, AK084160.1, AK140559.1, BC058413.1, NM_178777.3 transcripts.
Set Membership: |
|

Nhlh2
Genomic Map