Assay Details
| Gene Symbol: Serhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023475.3 | NP_075964.1 | 3 - 4 | 402 | 78 | |
| GenBank mRNA | AJ245737.1 | - | 405 | 78 | ||
| AJ251200.1 | - | 3 - 4 | 405 | 78 | ||
| AK002313.1 | - | 3 - 4 | 315 | 78 | ||
| AK003827.1 | - | 3 - 4 | 315 | 78 | ||
| AK154770.1 | - | 2 - 3 | 1610 | 78 | ||
| BC024048.1 | - | 3 - 4 | 283 | 78 | ||
| BC046342.1 | - | 3 - 4 | 287 | 6475904 | 78 | |
| BC055431.1 | - | 3 - 4 | 414 | 3673247 | 78 | |
| BC064069.1 | - | 2 - 3 | 472 | 78 | ||
More Information
Set Membership: |
|

Serhl
Genomic Map