Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 301 | Mir301 |
Mirn301, mir-301a, mmu-mir-301 |
-8.7kb | Anti-sense | Chr. 11 - 87113004 - 87113089 on NCBI Build GRCm38 |
| Downstream | tripartite motif-containing 37 | Trim37 |
1110032A10Rik, 2810004E07Rik, AI848587, AU043018, MUL, TEF3, mKIAA0898 |
3.2kb | Anti-sense | Chr. 11 - 87127051 - 87250874 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map