Assay Details
| Gene Symbol: Gm16907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045794.1 | 1 - 2 | 313 | 54 | ||
| GenBank mRNA | AK030196.1 | - | 1 - 2 | 314 | 54 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fructose bisphosphatase 1 | Fbp1 |
Fbp-2, Fbp2, Fbp3 |
-404.4kb | Sense | Chr. 13 - 62864753 - 62888296 on NCBI Build GRCm38 |
| Downstream | microRNA 23b | Mir23b |
Mirn23b, mmu-mir-23b |
3.3kb | Anti-sense | Chr. 13 - 63300484 - 63300557 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16907
Genomic Map